missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Based on Sepharose Fast Flow base matrix derivatized via uncharged, chemically-stable ether linkages
- Hydrophobic interaction chromatography (HIC) media composed of cross-linked 4% agarose
- Aliphatic butyl ligand immobilized to base matrix with ether linkage
- Fast Flow matrix gives the adsorbent high chemical, physical and thermal stabilities
- Dynamic capacity at low salt concentration
- Used with all aqueous buffers commonly used in chromatography
- Autoclavable at 121°C for 20 minutes in water
- Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
Specifications
Specifications
| Quantity | 200 mL |
| Stability | Stable in commonly used aqueous Buffers: 1 M NaOH, 30% Isopropanol, 70% Ethanol, 6 M Guanidine-Hydrochloride, 30% Acetonitrile, 1 mM HCl |
| For Use With (Application) | For Capture and Intermediate PurifiCatIon of Larger Proteins |
| Storage Requirements | 4°C to 30°C |
| Product Type | Butyl Sepharose 4 Fast Flow Media |
| Flow Rate | 150 to 250 cm/Hoμr |
| Particle Size | 45 to 165 μm |
| Height (English) Bed | 9.84 in. |
| Height (Metric) Bed | 25 cm |
| Max. Pressure | 100 kPa |
| Show More |
Safety and Handling
Product Identifier
- Butyl Sepharose 4 Fast Flow, 200 ml
- Warning
- Flammable liquid Category 3
- H302-Harmful if swallowed.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?