Learn More
Bupropion hydrochloride, 99%, Thermo Scientific™
A selective inhibitor of dopamine uptake
£43.50 - £124.00
Chemical Identifiers
| CAS | 31677-93-7 |
|---|---|
| Molecular Formula | C13H19Cl2NO |
| Molecular Weight (g/mol) | 276.201 |
| MDL Number | MFCD00055209 |
| InChI Key | HEYVINCGKDONRU-UHFFFAOYSA-N |
| Synonym | bupropion hydrochloride, wellbutrin, 2-tert-butylamino-1-3-chlorophenyl propan-1-one hydrochloride, bupropion hcl, zyban, wellbutrin xl, bupropion hydrocloride, wellbutrin sr, budeprion, forfivo xl |
| PubChem CID | 62884 |
| IUPAC Name | 2-(tert-butylamino)-1-(3-chlorophenyl)propan-1-one;hydrochloride |
| SMILES | CC(C(=O)C1=CC(=CC=C1)Cl)NC(C)(C)C.Cl |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15453759
|
Thermo Scientific Alfa Aesar
J61105.MD |
250mg |
£43.50
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15443759
|
Thermo Scientific Alfa Aesar
J61105.03 |
1g |
£124.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsBupropion HC is used as an inhibitor of dopamine and SLC6A2. Bupropion is efficacious as an antidepressant because of its effects on nicotinic receptors, it is also used to promote smoking cessation.
Solubility
Soluble in water (>25 mg/ml), ethanol 193mg/ml, 0.1 N HCl 333 mg/ml
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Chemical Identifiers
| 31677-93-7 | |
| 276.201 | |
| HEYVINCGKDONRU-UHFFFAOYSA-N | |
| 62884 | |
| CC(C(=O)C1=CC(=CC=C1)Cl)NC(C)(C)C.Cl |
| C13H19Cl2NO | |
| MFCD00055209 | |
| bupropion hydrochloride, wellbutrin, 2-tert-butylamino-1-3-chlorophenyl propan-1-one hydrochloride, bupropion hcl, zyban, wellbutrin xl, bupropion hydrocloride, wellbutrin sr, budeprion, forfivo xl | |
| 2-(tert-butylamino)-1-(3-chlorophenyl)propan-1-one;hydrochloride |
Specifications
| 31677-93-7 | |
| MFCD00055209 | |
| bupropion hydrochloride, wellbutrin, 2-tert-butylamino-1-3-chlorophenyl propan-1-one hydrochloride, bupropion hcl, zyban, wellbutrin xl, bupropion hydrocloride, wellbutrin sr, budeprion, forfivo xl | |
| HEYVINCGKDONRU-UHFFFAOYSA-N | |
| 2-(tert-butylamino)-1-(3-chlorophenyl)propan-1-one;hydrochloride | |
| 62884 | |
| 99% |
| C13H19Cl2NO | |
| 14,1499 | |
| Soluble in water (>25mg/ml),ethanol 193mg/ml,0.1 N HCl 333mg/ml | |
| CC(C(=O)C1=CC(=CC=C1)Cl)NC(C)(C)C.Cl | |
| 276.201 | |
| 276.2 | |
| Bupropion hydrochloride |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 250-759-9
missing translation for 'rtecsNumber' : UG8858000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.