missing translation for 'onlineSavingsMsg'
Learn More

Bupropion hydrochloride, 99%

Product Code. 15453759
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15453759 250 mg 250mg
15443759 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15453759 Supplier Thermo Scientific Alfa Aesar Supplier No. J61105.MD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A selective inhibitor of dopamine uptake | CAS: 31677-93-7 | C13H19Cl2NO | 276.201 g/mol

Bupropion HC is used as an inhibitor of dopamine and SLC6A2. Bupropion is efficacious as an antidepressant because of its effects on nicotinic receptors, it is also used to promote smoking cessation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bupropion HC is used as an inhibitor of dopamine and SLC6A2. Bupropion is efficacious as an antidepressant because of its effects on nicotinic receptors, it is also used to promote smoking cessation.

Solubility
Soluble in water (>25 mg/ml), ethanol 193mg/ml, 0.1 N HCl 333 mg/ml

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 31677-93-7
Molecular Formula C13H19Cl2NO
Molecular Weight (g/mol) 276.201
MDL Number MFCD00055209
InChI Key HEYVINCGKDONRU-UHFFFAOYSA-N
Synonym bupropion hydrochloride, wellbutrin, 2-tert-butylamino-1-3-chlorophenyl propan-1-one hydrochloride, bupropion hcl, zyban, wellbutrin xl, bupropion hydrocloride, wellbutrin sr, budeprion, forfivo xl
PubChem CID 62884
IUPAC Name 2-(tert-butylamino)-1-(3-chlorophenyl)propan-1-one;hydrochloride
SMILES CC(C(=O)C1=CC(=CC=C1)Cl)NC(C)(C)C.Cl

Spezifikation

Melting Point 233°C to 234°C
Color White
Quantity 250 mg
Merck Index 14,1499
Solubility Information Soluble in water (>25mg/ml),ethanol 193mg/ml,0.1 N HCl 333mg/ml
Formula Weight 276.2
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Bupropion hydrochloride

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.