missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Buffered Sodium Chloride Peptone 0.1% Tween

Product Code. 17870079
Click to view available options
Fill Weight:
500 to 504 g
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 17870079

Brand: Thermo Scientific™ BO0755V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recover and enumerate microorganisms from surfaces or products by neutralizing residual disinfectants and dispersing hydrophobic cells.

Recover and enumerate microorganisms from surfaces or products by neutralizing residual disinfectants and dispersing hydrophobic cells using Buffered Sodium Chloride Peptone 0.1% Tween.

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless, Clear
Content And Storage 2°C to 25°C
Format 500 mL Sirop Bottle
Manufacturing Origin United Kingdom
Product Type Microbiology Media
Fill Weight 500 to 504 g
Packaging Type 10 Sirop Bottles with screw cap closures in a box
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.