Learn More
Thermo Scientific™ Buffered Peptone Water (Bottled)
Description
Pre-enrich food samples prior to the isolation of Salmonella species using these ready-prepared media bottles.
Pre-enrich food samples prior to the isolation of Salmonella species with Thermo Scientific™ Oxoid™ Buffered Peptone Water ready-prepared bottled medium. Buffered Peptone Water is a non-selective pre-enrichment medium.
Low numbers and/or sub-lethally injured Salmonella cells may be present in a test food sample. Pre-enrichment in non-selective Buffered Peptone Water allows the cells time to repair and multiply before being introduced into selective culture, improving the chances of recovering Salmonella species from the sample.
- Convenient: the pre-filled bottles are ready for immediate use.
- Simple: easy-to-use with minimum training.
- Long shelf life: up to 12 months.
- Aids recovery of damaged Salmonella cells.
The test sample is added to Buffered Peptone Water at a ratio of 1:10 and incubated at 37° ± 1°C for 18 hours and 2 hours.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
Specifications
Specifications
| Color | Straw |
| Content And Storage | 2° to 25°C |
| Format | 125 mL Sirop Bottle |
| Manufacturing Origin | United Kingdom |
| Product Type | Microbiology Media |
| Fill Weight | 99 to 101 g |
| Packaging Type | 10 Sirop Bottles with screw cap closures in a box |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.