missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Buffered Peptone Water

Product Code. 15842357
Change view
Click to view available options
Fill Weight:
223-227 g
950 to 955 g
Format:
1 L Sirop Bottle
225 mL sirop bottle
Unit Size:
1 set
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight Format unitSize
15842357 950 to 955 g 1 L Sirop Bottle 1 set
12635885 223-227 g 225 mL sirop bottle Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15842357 Supplier Thermo Scientific™ Supplier No. EB1067Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pre-enrich food samples prior to the isolation of Salmonella species with this non-selective medium while adhering to ISO 11133:2014.

Pre-enrich food samples prior to the isolation of Salmonella species with Buffered Peptone Water.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw
Content And Storage 2°C to 25°C
Format 1 L Sirop Bottle
Manufacturing Origin United Kingdom
Product Type Microbiology Media
Fill Weight 950 to 955 g
Packaging Type 4 Sirop Bottles with Screw Cap Closures/Box
Quantity 4/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.