missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Buffered Peptone Water
Click to view available options
Fill Weight:
223-227 g
950 to 955 g
Format:
1 L Sirop Bottle
225 mL sirop bottle
Unit Size:
1 set
Pack of 10
Description
Pre-enrich food samples prior to the isolation of Salmonella species with Buffered Peptone Water.
Specifications
Specifications
| Color | Straw |
| Format | 225 mL sirop bottle |
| Fill Weight | 223-227 g |
| Packaging Type | 10 sirop bottles with screw cap closure in a box |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction