missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Buffered Peptone Water

Product Code. 15802357
Change view
Click to view available options
Quantity:
4 x 1000 mL
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15802357 4 x 1000 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15802357 Supplier Thermo Scientific™ Supplier No. EB0201W

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

BUFFERED PEPTONE WATER 4 x 1000 mL: A non-selective pre-enrichment medium for the detection of Salmonella, Listeria and Enterobacteriaceae from food and water samples

A non-selective pre-enrichment medium for the detection of Salmonella, Listeria and Enterobacteriaceae from food and water samples

  • A non-selective pre-enrichment medium for the detection of Salmonella, Listeria and Enterobacteriaceae from food and water samples
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Manufacturing Origin United Kingdom
Product Type Broth Tube
Sterility Non-sterile
Target Organism Class Bacterial Pathogens
Test Type Microbial Detection
Quantity 4 x 1000 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.