missing translation for 'onlineSavingsMsg'
Learn More

BTC Mouse anti-Human, Clone: 2H5E8, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen BTC
Clone 2H5E8
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16828795
View Documents
Proteintech
66683-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16818795
View Documents
Proteintech
66683-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The protein encoded by this gene is a member of the EGF family of growth factors. It is synthesized primarily as a transmembrane precursor, which is then processed to mature molecule by proteolytic events. This protein is a ligand for the EGF receptor.
Specifications

Specifications

BTC
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
betacellulin, BTC, Probetacellulin
BTC
IgG1
Protein G
Antibody
2H5E8
Western Blot
Unconjugated
Mouse
Human
P35070
685
BTC Fusion Protein Ag26724
Primary
-20°C
BTC
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.