missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTC Mouse anti-Human, Clone: 2H5E8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66683-1-IG-20UL
This item is not returnable.
View return policy
Beschreibung
The protein encoded by this gene is a member of the EGF family of growth factors. It is synthesized primarily as a transmembrane precursor, which is then processed to mature molecule by proteolytic events. This protein is a ligand for the EGF receptor.Spezifikation
| BTC | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P35070 | |
| BTC | |
| BTC Fusion Protein Ag26724 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 2H5E8 | |
| Unconjugated | |
| BTC | |
| betacellulin, BTC, Probetacellulin | |
| Mouse | |
| Protein G | |
| RUO | |
| 685 | |
| -20°C | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur