missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ BTC, AM, cell permeant

Product Code. 11544197
Click to view available options
Quantity:
100 μg
Unit Size:
1000µg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11544197

Brand: Molecular Probes™ B6791

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

BTC, AM, cell permeant

BTC, AM is a cell-permeant coumarin benzothiazole-based Ca2+ indicator. It exhibits a shift in excitation maximum from about 480 nm to 400 nm upon binding Ca2+, which permits ratiometric measurements that are essentially independent of uneven dye loading cell thickness, photobleaching and dye leakage. BTC has high selectivity for Ca2+ and a Kd ∼7μM enabling quantitation of high intracellular Ca2+ levels.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Cell Viability and Proliferation
For Use With (Equipment) Flow Cytometer
Product Type Calcium Indicator
Quantity 100 μg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.