missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTBD8 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | BTBD8 |
|---|---|
| Concentration | 0.43 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16882505
|
Proteintech
23897-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16872505
|
Proteintech
23897-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BTBD8, also named as BTB/POZ domain-containing protein 8, is a 378 amino acid protein, which is highly expressed in fetal brain and localizes in the nucleus. BTBD8 exists two isoforms and the molecular weight of isoforms are 43 kDa and 35 kDa.Specifications
| BTBD8 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q5XKL5 | |
| 284697 | |
| BTBD8 Fusion Protein Ag20960 | |
| Primary | |
| -20°C | |
| BTBD8 |
| 0.43 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| BTB (POZ) domain containing 8, BTBD8 | |
| BTBD8 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title