missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTBD8 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23897-1-AP-150UL
This item is not returnable.
View return policy
Description
BTBD8, also named as BTB/POZ domain-containing protein 8, is a 378 amino acid protein, which is highly expressed in fetal brain and localizes in the nucleus. BTBD8 exists two isoforms and the molecular weight of isoforms are 43 kDa and 35 kDa.Specifications
| BTBD8 | |
| Polyclonal | |
| Unconjugated | |
| BTBD8 | |
| BTB (POZ) domain containing 8, BTBD8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 284697 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5XKL5 | |
| BTBD8 | |
| BTBD8 Fusion Protein Ag20960 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction