Learn More
Bronze gauze, alloy 220, 200 mesh woven from 0.053mm (0.0021in) in dia wire, Thermo Scientific™
£20.50 - £69.60
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15466835
|
Thermo Scientific Alfa Aesar
045223.HB |
300 x 300 mm |
£69.60
Each |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15456835
|
Thermo Scientific Alfa Aesar
045223.GT |
150 x 150 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||