missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Bromocresol Purple Agar

Product Code. 13295359
Change view
Click to view available options
Quantity:
10 Plates
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13295359 10 Plates Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13295359 Supplier Thermo Scientific™ Supplier No. PO5118A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Optimize bacterial isolation with this agar, providing clear differentiation and reliable results.

Optimize bacterial isolation with Bromocresol Purple Agar, providing clear differentiation and reliable results.

TRUSTED_SUSTAINABILITY

Specifications

Color Transparent
Content And Storage 2°C to 12°C
Format 90 mm
Manufacturing Origin Germany
Product Type Agar Plate
Fill Volume 17 mL
Quantity 10 Plates
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.