missing translation for 'onlineSavingsMsg'
Learn More

Bromobenzene, 99%

Product Code. 11440243
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
This item is not returnable. View return policy

Product Code. 11440243

Brand: Thermo Scientific Alfa Aesar A11407.0C

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-86-1 | C6H5Br | 157.01 g/mol

Bromobenzene is used as solvent in organic synthesis. It is used to prepare the phenylmagnesium bromide, a Grignard reagent, an intermediate in the preparation of benzoic acid by reacting with carbon dioxide. It is a precursor for making phencyclidine. It is useful for the introduction of phenyl group through palladium-catalyzed coupling reactions viz. the Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromobenzene is used as solvent in organic synthesis. It is used to prepare the phenylmagnesium bromide, a Grignard reagent, an intermediate in the preparation of benzoic acid by reacting with carbon dioxide. It is a precursor for making phencyclidine. It is useful for the introduction of phenyl group through palladium-catalyzed coupling reactions viz. the Suzuki reaction.

Solubility
Miscible with diethyl ether, alcohol, carbon tetrachloride, chloroform and benzene.

Notes
Store in cool place. Avoid inhalation of vapor or mist. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-86-1
Molecular Formula C6H5Br
Molecular Weight (g/mol) 157.01
MDL Number MFCD00000055
InChI Key QARVLSVVCXYDNA-UHFFFAOYSA-N
Synonym benzene, bromo, monobromobenzene, phenyl bromide, bromo-benzene, 1-bromobenzene, phbr, bromo benzene, 4-bromobenzene, unii-co4d5j547l, hsdb 47
PubChem CID 7961
ChEBI CHEBI:3179
IUPAC Name bromobenzene
SMILES BrC1=CC=CC=C1

Specifications

Melting Point -31°C
Density 1.492
Boiling Point 155°C to 156°C
Flash Point 51°C (123°F)
Assay Percent Range 99%
Odor Aromatic
Refractive Index 1.559
Quantity 10,000 g
UN Number UN2514
Beilstein 1236661
Merck Index 14,1406
Solubility Information Miscible with diethyl ether,alcohol,carbon tetrachloride,chloroform and benzene.
Formula Weight 157.02
Percent Purity 99%
Chemical Name or Material Bromobenzene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bromobenzene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P362-Take off contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contains : Bromobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.