missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzyl bromide, 99+%, Thermo Scientific™

Product Code. 10417540 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10417540 100 g Plastic bottle 100g
10776171 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10417540 Supplier Thermo Scientific Acros Supplier No. 146131000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20443-98-5
Molecular Formula C7H5BrCl2
Molecular Weight (g/mol) 239.92
MDL Number MFCD00000577
InChI Key PDFGFQUSSYSWNI-UHFFFAOYSA-N
Synonym 2,6-dichlorobenzyl bromide, 2-bromomethyl-1,3-dichlorobenzene, alpha-bromo-2,6-dichlorotoluene, 2,6-dichlorobenzylbromide, benzene, 2-bromomethyl-1,3-dichloro, 1-bromomethyl-2,6-dichlorobenzene, .alpha.-bromo-2,6-dichlorotoluene, a-bromo-2,6-dichlorotoluene, 2,6 dichloro benzyl bromide, benzene, 2-bromomethyl-1,3-dichloro-9ci
PubChem CID 30159
IUPAC Name 2-(bromomethyl)-1,3-dichlorobenzene
SMILES ClC1=CC=CC(Cl)=C1CBr

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 54°C to 57°C
Color Colorless to White
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula Cl2C6H3CH2Br
Quantity 100 g
Solubility Information Solubility in water: .. Other solubilities: 1g/10mL methanol
Formula Weight 239.93
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2, 6-Dichlorobenzyl bromide, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 2,6-Dichlorobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.