accessibility menu, dialog, popup

UserName

ClClBr

CAS RN 20443-98-5

IUPAC Name: 2-(bromomethyl)-1,3-dichlorobenzene
Synonyms: 2-(bromomethyl)-1,3-dichlorobenzene
Molecular Weight (g/mol): 239.92
Molecular Formula: C7H5BrCl2
InChi Key: PDFGFQUSSYSWNI-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(Cl)=C1CBr
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)