missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BRMS1 Recombinant Rabbit Monoclonal Antibody (23GB3015), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | BRMS1 |
|---|---|
| Clone | 23GB3015 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087681
![]() |
Invitrogen™
MA553312 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088395
|
Invitrogen™
MA553313 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BRMS1 Recombinant Monoclonal antibody specifically detects BRMS1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| BRMS1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9HCU9 | |
| 25855 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3015 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AV003220; AW554636; breast cancer metastasis suppressor 1; breast cancer metastasis-suppressor 1; Breast cancer metastasis-suppressor 1 homolog; BRMS 1; BRMS1 | |
| A synthesized peptide derived from BRMS1 (1-56AA). | |
| Primary | |
| -20°C | |
| BRMS1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title