missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BRMS1 Recombinant Rabbit Monoclonal Antibody (23GB3015), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553312
This item is not returnable.
View return policy
Description
This gene reduces the metastatic potential, but not the tumorogenicity, of human breast cancer and melanoma cell lines. The protein encoded by this gene localizes primarily to the nucleus and is a component of the mSin3a family of histone deacetylase complexes (HDAC). The protein contains two coiled-coil motifs and several imperfect leucine zipper motifs. Alternative splicing results in two transcript variants encoding different isoforms.
Specifications
| BRMS1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| BRMS1 | |
| AV003220; AW554636; breast cancer metastasis suppressor 1; breast cancer metastasis-suppressor 1; Breast cancer metastasis-suppressor 1 homolog; BRMS 1; BRMS1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 25855 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3015 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9HCU9 | |
| BRMS1 | |
| A synthesized peptide derived from BRMS1 (1-56AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction