missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brk Antibody (2H12B8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60537-0.025ml
This item is not returnable.
View return policy
Description
Brk Monoclonal specifically detects Brk in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Brk | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| Breast tumor kinase, BRKprotein-tyrosine kinase 6, EC 2.7.10, EC 2.7.10.2, FLJ42088, protein-tyrosine kinase BRK, PTK6 protein tyrosine kinase 6, Tyrosine-protein kinase BRK | |
| Mouse | |
| 52 kDa | |
| 0.025 mL | |
| Protein Kinase | |
| 5753 | |
| Human | |
| Purified |
| Western Blot | |
| 2H12B8 | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| PTK6 | |
| Ni-NTA purified truncated recombinant PTK6 expressed in E. Coli strain BL21 (DE3). | |
| Unpurified | |
| RUO | |
| Primary | |
| PTK6 (2H12B8) | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2b Kappa |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction