missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brk Antibody (2H12B8), Novus Biologicals™
Mouse Monoclonal Antibody
£157.00
Specifications
| Antigen | Brk |
|---|---|
| Clone | 2H12B8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
| Applications | Western Blot |
Description
Brk Monoclonal specifically detects Brk in Human samples. It is validated for Western Blot, ELISA.Specifications
| Brk | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Protein Kinase | |
| Ascites with 0.03% Sodium Azide | |
| 5753 | |
| Ni-NTA purified truncated recombinant PTK6 expressed in E. Coli strain BL21 (DE3). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 52 kDa |
| 2H12B8 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Breast tumor kinase, BRKprotein-tyrosine kinase 6, EC 2.7.10, EC 2.7.10.2, FLJ42088, protein-tyrosine kinase BRK, PTK6 protein tyrosine kinase 6, Tyrosine-protein kinase BRK | |
| PTK6 | |
| IgG2b Kappa | |
| Unpurified | |
| PTK6 (2H12B8) |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title