missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRIP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£133.90 - £349.17
Specifications
| Antigen | BRIP1 |
|---|---|
| Concentration | 0.73 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
The antibody is specific to BRIP1.
LIN54 is a component of the LIN, or DREAM, complex, an essential regulator of cell cycle genes.Specifications
| BRIP1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9BX63 | |
| 83990 | |
| Peptide | |
| Primary | |
| -20°C | |
| BRIP1 |
| 0.73 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| BACH1, BRIP1, FANCJ, OF | |
| BRIP1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title