missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRIP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20983-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to BRIP1.
LIN54 is a component of the LIN, or DREAM, complex, an essential regulator of cell cycle genes.Specifications
| BRIP1 | |
| Polyclonal | |
| Unconjugated | |
| BRIP1 | |
| BACH1, BRIP1, FANCJ, OF | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 83990 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.73 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BX63 | |
| BRIP1 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction