Learn More
Thermo Scientific™ Brillliance™ VRE Agar
Description
Improve the prevention and control of VRE in healthcare settings by rapidly and reliably isolating and presumptively identifying vancomycin-resistant Enterococcus (VRE) within 24 hours direct from sample using Thermo Scientific™ Brilliance™ VRE. Vancomycin-resistant Enterococcus faecium and Enterococcus faecalis are differentiated through the utilization of two separate chromogens. Interpretation is aided through the inclusion of a semi-opaque background, providing a distinct visual contrast against light blue and indigo colonies.
Additional antibiotics, in combination with vancomycin, are present to suppress the growth of competing flora including E. gallinarum and E. casseliflavus, both of which are intrinsically resistant to vancomycin, possessing the chromosomally encoded VanC resistance mechanism. The VanC resistance mechanism is not readily transmissible between organisms and, as such, is deemed less clinically significant than VanA and VanB mechanisms which are encoded on freely transmissible genetic elements, plasmids and transposons, thus increasing the risk of resistance genes spreading to other organisms.
- Rapidly and economically detects colonization of Vancomycin-Resistant Enterococcus (VRE) colonization
- Aids in the prevention and controls the spread of vancomycin resistance in healthcare settings
- Correctly classifies patients whether positive or negative
- Provides superior results in a selective and differential chromogenic medium
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Color | Pale buff, opaque |
| Content And Storage | 2°C to 10°C |
| Description | Brilliance VRE Agar |
| Format | 90mm Monoplate |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar Plate |
| Fill Weight | 19.5g ± 1.0g |
| Packaging Type | Flow Wrapped - Single Layer |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.