missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brilliance™ ESBL Agar/ Brilliance™ CRE Agar

Product Code. 15832447
Change view
Click to view available options
Description:
Brilliance ESBL Agar/ Brilliance CRE Agar
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Quantity unitSize
15832447 Brilliance ESBL Agar/ Brilliance CRE Agar 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15832447 Supplier Thermo Scientific™ Supplier No. PO1265E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Detect and identify ESBL- and CRE-producing bacteria in clinical samples accurately, easily and quickly with this chromogenic, bi-plate combination.

Detect CRE-and ESBL-colonized patients accurately, easily and quickly with Thermo Scientific™ Brilliance™ ESBL Agar/ Brilliance™ CRE Agar chromogenic bi-plates. Screen clinical samples simultaneously for both extended spectrum β-lactamase- (ESBL) producing organisms and carbapenem-resistant Enterobacteriaceae (CRE) on a single plate for more timely and targeted treatment of patients.

Screen clinical samples faster with Brilliance ESBL Agar/ Brilliance CRE Agar bi-plates which combine two easy-to-read, chromogenic selective media. The bi-plate combination allows the easy identification of clinically relevant ESBL-producing organisms and CRE.

  • Easily and clearly differentiate E.coli and the Klebsiella, Enterobacter, Serratia and Citrobacter (KESC) group by different colors
  • Reduce false positives and the need for confirmatory tests through inhibition of AmpC producers
  • Achieve results in just 18 hours for carbapenem-resistant Enterobacteriaceae (CRE)

Brilliance ESBL Agar contains cefpodoxime, in combination with additional antibacterial agents, to inhibit non-ESBL Enterobacteriaceae and to suppress the growth of most AmpC organisms and other non-ESBL flora.

Differentiation of the most prevalent ESBL-producing organisms is achieved through the inclusion of two chromogens that specifically target two enzymes. Members of the KESC group express galactosidase, resulting in green colonies. E. coli, however, express galactosidase and glucuronidase which, depending on the activity of the respective enzymes, will appear as blue colonies. Galactosidase negative E. coli (less than 5%) will appear pink. Proteus, Morganella and Providencia do not utilise either chromogen but are able to deaminate tryptohan, resulting in tan-coloured colonies with a brown halo. Colorless colonies refer to Salmonella, Acinetobacter or other bacteria with other resistance mechanisms which may grow on this medium. Any such isolate may be clinically significant and should be investigated further.

Brilliance CRE Agar contains a modified carbapenem, at a level recommended by both EUCAST and CLSI, ensuring reliable results with a wide variety of CRE, including those with the New Delhi Metallo-β-lactamase-1 (NDM-1) mechanism. The two-chromogen system differentiates E. coli (pink) from the KESC group, which grow blue. The novel background makes the white or naturally pigmented colonies of non-CRE organisms, such as Acinetobacter, easy-to-spot, and results are available in just 18 hours.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 10°C
Description Brilliance ESBL Agar/ Brilliance CRE Agar
Format 90mm bi-plate
Product Type Agar Plate
Fill Weight 19.5g ± 1g
Packaging Type Wrapped in single cellulose-based film wrap
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.