missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brilliance™ E. coli / Coliform Selective Agar

Product Code. 11964102
Change view
Click to view available options
Description:
Brilliance E. coli / Coliform Selective
Quantity:
10/Pk.
Fill Volume:
17 mL
Manufacturing Origin:
Germany
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Quantity Fill Volume Manufacturing Origin Unit Size
11964102 Brilliance E. coli / Coliform Selective 10/Pk. 17 mL Germany Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11964102 Supplier Thermo Scientific™ Supplier No. PO5176A
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detect and enumerate Escherichia coli and other coliforms in food and water samples with this selective, chromogenic medium.

Detect and enumerate Escherichia coli and other coliforms in food and water samples with selective, chromogenic Thermo Scientific™ Brilliance™ E. coli / Coliform Selective Agar. The agar base uses two chromogens to differentiate between Escherichia coli and other coliforms. This results in purple E. coli, as they are able to cleave both chromogens with other coliforms, giving pink colonies.

The recovery and enumeration of Escherichia coli and coliforms are important indicators of environmental and food hygiene. Brilliance E. coli / Coliform Selective Agar contains two chromogenic agents:

  • Rose-Gal: detects ß-galactosidase activity
  • X-Glu: detects ß-glucuronidase activity

Detection of ß-glucuronidase activity is widely used to differentiate Escherichia coli, as this is present in E. coli but not in other members of the coliform group. As most coliforms are lactose-positive, ß -galactosidase activity, encoded by the lacZ gene, is then used to differentiate this group from other organisms able to grow on the selective medium. This results in purple E. coli, as they are able to cleave both chromogens with other coliforms, giving pink colonies, as they cleave only the galactoside chromogen.

The medium also contains sodium lauryl sulphate which acts as a selective agent, inhibiting the growth of Gram-positive organisms. Due to the addition of tryptophan, the rapid confirmation of E. coli by the formation of indole using a spot indole test is possible.
 

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Color Ivory, transparent
Content And Storage 2° to 12°C, in the dark
Description Brilliance E. coli / Coliform Selective
Format 90mm Monoplate
Manufacturing Origin Germany
Product Type Agar Plate
Fill Volume 17 mL
Fill Weight 17g ± 5%
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.

For laboratory use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.