missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brilliance™ CRE Agar

Product Code. 13205349
Change view
Click to view available options
Format:
90mm Monoplate
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Unit Size
13205349 90mm Monoplate Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13205349 Supplier Thermo Scientific™ Supplier No. PO1226A
Spotlight Offers
Use promo code 28038
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detect CRE-colonized patients easier and earlier with a chromogenic screening plate that provides results for carbapenem-resistant Enterobacteriaceae (CRE) in just 18 hours.

Detect CRE-colonized patients easier and earlier using Thermo Scientific™ Brilliance™ CRE Agar, a chromogenic screening plate that provides results in just 18 hours for carbapenem-resistant Enterobacteriaceae (CRE). Allow for more timely and targeted treatment of patients while limiting the potential transmission and spread of these pathogenic organisms.

Traditionally, CRE detection has relied on antimicrobial susceptibility testing in conjunction with synergistic methods, like the Modified Hodge Test (MHT). However, MHT is technically demanding, and the results are often subjective and open to interpretation, so is no longer recommended by the CLSI or EUCAST. As the number of CRE being isolated has increased, especially in geographies such as India, Greece, Israel, Turkey and New York in the U.S.A., so has the need for an accurate, easy-to-use screening method to help identify colonized patients more quickly.

Brilliance CRE Agar makes detecting CRE-colonized patients easy. The formulation contains a modified carbapenem at a level recommended by both EUCAST and CLSI, ensuring reliable results with a wide variety of carbapenem-resistant organisms, including New Delhi Mettalo β-lactamase -1 (NDM-1). The two-chromogen system differentiates E. coli (pale pink) from the KESC group organisms, which grow blue. Other carbapenem-resistant organisms not from the Enterobacteriaceae family, e.g., Acinetobacter, may grow, but they produce white or naturally pigmented colonies which are easy to spot against the novel turquoise background. The novel background makes the white or naturally pigmented colonies of non-CRE organisms, like Acinetobacter, easy-to-spot, all of which means you can screen for CRE direct from patient samples in just 18 hours.

Plates are incubated at 37ºC and provide high sensitivity and specificity with results available in just 18 to 24 hours. This allows a rapid response from infection control teams and enables the patient to receive the most appropriate treatment as early as possible. Accuracy minimizes costs by ensuring that only those in need receive what can be costly treatment.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Color Blue/ Green, opaque
Content And Storage 2°C to 10°C
Description Brilliance CRE Agar
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Product Type Agar Plate
Fill Weight 19.5g ± 1.0g
Packaging Type Flow wrapped - single layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.