missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brilliance™ Candida Agar / Sabouraud GC Agar

Product Code. 15853247
This item is not returnable. View return policy

Product Code. 15853247

Brand: Thermo Scientific™ PO5258E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate fungi and differentiate and presumptively identify Candida albicans and Candida tropicalis from clinical specimens with this bi-plate.

Isolate fungi and differentiate and presumptively identify Candida albicans and Candida tropicalis from clinical specimens, with ready-to-use Thermo Scientific™ Brilliance™ Candida Agar / Sabouraud GC Agar, a single bi-plate which includes two separate formulations. The Sabouraud GC medium creates selective conditions for isolation and cultivation of fungi while Brilliance™ Candida Agar enables the isolation and differentiation of Candida albicans and Candida tropicalis from other Candida spp. within 48 hours. The convenient bi-plate format frees up incubation space.

Use Brilliance™ Candida Agar / Sabouraud GC Agar for isolation of fungi and presumptive identification Candida albicans and Candida tropicalis from other Candida spp. from clinical specimens.

  • Suitable for identification of pathogenic fungi from material containing large numbers of other fungi or bacteria
  • Easy isolation, differentiation and presumptive identification of Candida albicans and Candida tropicalis from other Candida within 48 hours.
  • Ready-to-use convenience of prepared media.
  • Space saving bi-plate format.

Brilliance Candida Agar incorporates two chromogens that indicate the presence of the target enzymes. As a result, Candida tropicalis form dark blue colonies while Candida albicans form Green colored colonies.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Oyster white / Ivory
Content And Storage 2° to 12°C, in the dark
Format 90mm Biplate
Manufacturing Origin Germany
Product Type Agar
Fill Weight 17g ± 5%
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.