missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRE Mouse anti-Human, Mouse, Rat, Clone: 2E5A5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66356-1-IG-150UL
This item is not returnable.
View return policy
Description
BRCC45 was initially suggested to be a housekeeping protein that is highly expressed in brain and reproductive organs. Later experiments indicated BRCC45 forms a complex with the breast and ovarian predisposition proteins BRCA1 and BRCA2 as well as RAD51 and BRCC36. This complex has a ubiquitin E3 ligase activity and is thought to enhance cellular survival following DNA damage. BRCC45 has also been suggested to function as a death receptor-associated anti-apoptotic protein by inhibiting the BID-induced activation of the mitochondrial apoptotic pathway. Higher levels of BRCC45 were detected in the majority of hepatocellular carcinomas, suggesting that BRCC45 may promote tumorigenesis when overexpressed. At least three isoforms of BRCC45 are known to exist.Specifications
| BRE | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6P7Q1, Q8K3W0, Q9NXR7 | |
| Babam2 | |
| BRE Fusion Protein Ag2256 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 2E5A5 | |
| Unconjugated | |
| BABAM2 | |
| BRCA1 A complex subunit BRE, BRCC4, BRCC45, BRE | |
| Mouse | |
| Protein A | |
| RUO | |
| 107976, 362704, 9577 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction