Learn More
Thermo Scientific™ Brazier's Clostridium Difficile Selective Agar (HPA)
Description
Isolate and presumptively identify Clostridium difficile from clinical specimens with Thermo Scientific™ Oxoid™ Brazier's Clostridium Difficile Selective Agar (HPA). The presence of p-hydroxyphenylacetic acid in the medium promotes the production of p-cresol - a metabolite of C. difficile that produces a distinct odour to help in the presumptive identification. The addition of selective agents D-cycloserine and cefoxitin inhibit many bacteria that would normally be present in large numbers within fecal samples such as Enterobacteriaceae and Enterococcus faecalis.
Clostridium difficile was first isolated in 1935 by Hall and O’Toole1 who proposed the name ‘difficile’ because it was very difficult to isolate. Toxicogenic isolates of C. difficile have been demonstrated to be a major cause of antibiotic-associated ileo-caecitis in laboratory animals2 and pseudomembranous colitis in man3,4
Use Brazier’s Clostridium Difficile Selective Agar for the isolation and presumptive identification of Clostridium difficile from clinical samples.
- Helps presumptive identification of difficile through odour detection of p-cresol.
- Selects for C. difficile from the abundant flora that can be present within fecal samples.
- Easily differentiates difficile from lecithinase positive clostridia due to addition of Egg Yolk Emulsion. Lecithinase breaks down lecithin present in egg yolk emulsion producing insoluble opaque precipitation around the colony.
- Optimises UV light recognition of colonies due to the presence of lysed horse blood.
- Ready-to-use convenience of prepared media
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Color | Amber/Red |
| Content And Storage | 2°C to 10°C |
| Description | Brazier's Clostridium Difficile Selective Agar (HPA) |
| Format | 90mm Monoplate |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar Plate |
| Fill Weight | 19.5g±1.0.g |
| Packaging Type | 10 x 90mm plates, film wrapped |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.