missing translation for 'onlineSavingsMsg'
Learn More

R&L Enterprises™ Meker Brass Bunsen Burner with Gas Cartridge

Product Code. 12802154 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
200 g
Unit Size:
0.2kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12802154 200 g 0.2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12802154
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An excellent solution for heating, sterilization and combustion in the laboratory. R&L Enterprises™ Meker Brass Bunsen Burners with Gas Cartridge are designed for use with butane/propane mix gas cartridges.

  • Achieves temperatures in excess of 1000°C
  • Design of gas cartridge enables the burner to be detached and reattached as often as needed, making it easy to store and transport
  • Entirely constructed in brass with a replaceable steel perforated head
  • When used with the 220g cartridge the burner fits the standard 200mm-high tripod
  • Typical burning time of the 220g cartridge is 2.75hr

Specifications

Product Type Bunsen Burner
For Use With (Equipment) Butane/Propane Mix Gas Cartridges
Material Brass, Steel
Capacity (Metric) 200 g
Includes 220 g Cartridge
compliance-icons
Product Identifier
  • Butan
Signal Word
  • Danger
Hazard Category
  • Flammable gas.
Hazard Statement
  • H220-Extremely flammable gas.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P377-Leaking gas fire: Do not extinguish, unless leak can be stopped safely.
  • P381-Eliminate all ignition sources if safe to do so.
  • P403-Store in a well-ventilated place.
Supplemental information
  • MIXTURE LIST-Contain: Hydrocarbon C3-C4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.