Learn More
BrandTech™ VACUUBRAND™ VACUUBUS™ CVC3000 Vacuum Controller Solenoid Valves
For use with CVC3000 Vacuum Controller
£265.00 - £610.00
Products 3
Product Code Brand Type For Use With Price Quantity & Availability  
Product Code Brand Type For Use With Price Quantity & Availability  
10681284
View Documents Promotion Details
VACUUBRAND™
20674291
Solenoid Isolation Valve CVC3000 vacuum controller and DCP3000 vacuum gauge
£610.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
10353863
View Documents Promotion Details
VACUUBRAND™
20674220
Solenoid Coolant Valve CVC3000 vacuum controller
£265.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Add to basket
10099052
View Documents Promotion Details
VACUUBRAND™
20674217
Solenoid Vent Valve CVC3000 vacuum controller
£294.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
Description
Description
  • State-of-the-art vacuum gauges for laboratory and industrial operation have to meet high standards in terms of gauge head resistance to chemicals, reliability in rugged applications and operator-friendly interfaces. VACUUBRAND has a comprehensive range of electronic measuring instruments for the range of atmospheric pressure to 5 x 10-9 mbar.
  • by cyclically turning the pump on/off (with CVC 3000 controller with module VMS-B)
  • by cyclically opening/closing an in-line solenoid valve (CVC 3000) or
  • by changing the pump´s motor speed continuously (VARIO™ pumps in connection with CVC 3000)