Learn More
BRANDTECH™ VACUUBRAND™ Service Kits for MZ 2C Vacuum Pumps
Product Code. 10304351 Shop All Vacuubrand Products
Change view
Click to view available options
Type:
Service Kit
Unit Size:
1 set
Product Code. Type unitSize
10304351 Service Kit 1 set
1 options
Product Code. 10304351 Supplier BRANDTECH™ VACUUBRAND™ Supplier No. 20696814
Only null left
Add to basket
Add to basket
Service kits for VACUUBRAND™ Chemistry Diaphragm Pumps.
  • Sets include appropriate diaphragms, valves and sealing rings (if applicable)
  • Wrench not included.
TRUSTED_SUSTAINABILITY
Specifications
For Use With Model ME4C and MZ2C vacuum pumps
Includes 2 Diaphragms and 4 valves
Type Service Kit