Learn More
VACUUBRAND™ Rotary Vane Vacuum Pumps: Oil Mist Filters
Product Code. 10590362 Shop All Vacuubrand Products
Change view
Click to view available options
Type:
Filter Element
Oil Mist Filter
For Use With:
RE/RZ 2, 2.5, 5, 6 Rotary Vane Pumps
RE/RZ 8, 9, 16 Rotary Vane Pumps
RZ 2.5, RZ 6 Oil Mist Filter
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With unitSize
10590362 Filter Element RZ 2.5, RZ 6 Oil Mist Filter Each
10292551 Oil Mist Filter RE/RZ 2, 2.5, 5, 6 Rotary Vane Pumps Each
10485823 Oil Mist Filter RE/RZ 8, 9, 16 Rotary Vane Pumps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10590362 Supplier VACUUBRAND™ Supplier No. 20698004
Only null left
Add to basket
Add to basket
Oil Mist Filers and replacements for VACUUBRAND™ Rotary Vane Vacuum Pumps
  • For use with VACUUBRAND™ Rotary Vane Vacuum Pumps
  • Oil Mist Filters 'FO' trap up to 99% of oil mist generated by the pump, helping preserve the laboratory environment
TRUSTED_SUSTAINABILITY
Specifications
For Use With RZ 2.5, RZ 6 Oil Mist Filter
Type Filter Element