Learn More
VACUUBRAND™ Rotary Vane Vacuum Pumps: Inlet Separators
Product Code. 10652222 Shop All Vacuubrand Products
Change view
Click to view available options
For Use With:
RE2, RZ2, RE 2.5, RZ 2.5
RE5, RZ5, RE6, RZ6, RE 2.5, RZ 2.5
RE8, RE9, RE16, RZ8, RZ9, RZ16
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
10652222 RE2, RZ2, RE 2.5, RZ 2.5 Each
10394391 RE5, RZ5, RE6, RZ6, RE 2.5, RZ 2.5 Each
10707071 RE8, RE9, RE16, RZ8, RZ9, RZ16 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10652222 Supplier VACUUBRAND™ Supplier No. 20698000
Only null left
Add to basket
Add to basket
Inlet separators for VACUUBRAND™ Rotary Vane Vacuum Pumps.
  • For use with VACUUBRAND™ Rotary Vane Vacuum Pumps
  • Inlet Separators 'AK' help keep condensates and particulates from entering the vacuum pump, preserving performance.
Note: For RE/RZ 2.5: For rotary vane pumps RE 2.5 (article no. 2079715x) and RZ 2.5 (article no. 2079812x) - please order Inlet Separator 20698006. For rotary vane pumps RE 2.5 (article no. 2069715x) and RZ 2.5 (article no. 2069812x) - please order Inlet Separator 20698000.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) To keep condensates and particulates from entering the vacuum pump
For Use With RE2, RZ2, RE 2.5, RZ 2.5
Model 20698000
Type Inlet Catchpot/Separator