missing translation for 'onlineSavingsMsg'
Learn More

BrandTech™ Transferpette™ S Single Channel Fixed Volume Pipet

Uses innovative and ergonomic one-handed operation. BrandTech™ Transferpette™ S Single Channel Fixed Volume Pipet offers robust yet lightweight construction. Easy Calibration™ technology provides long lasting reliability, accuracy, and precision.

Brand:  BrandTech™ 704738

Code : 03

Additional Details : Weight : 0.31200kg

 View more versions of this product

Product Code. 10695453

  • £172.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Device is lightweight with one-handed volume adjustment.

  • Centralized pipetting button with optimized stroke length
  • Easy in-lab service
  • Robust and lightweight
  • Accepts universal tips
  • Autoclavable at 121°C/250°F (20 minutes)

Specifications

Specifications

±0.6%
ISO9001, CE, IVD, certificate of performance, individual calibration certificate
2 to 200μL tip, 5 to 300μL tip, 5 to 100μL tip
3 Years
100μL
Yes
2 to 200μL tip, 5 to 300μL tip, 5 to 100μL tip
Centralized piston button design, shelf mount, 4 digit display, steam sterilizable shaft, operating manual
F100
Videos
Documentation View accessory products