Learn More
VACUUBRAND™ BVC Shuttle for BVC Fluid Aspiration Systems
Product Code. 15860660 Shop All Vacuubrand Products
Click to view available options
For Use With:
BVC Fluid Aspiration System
Unit Size:
Each
Product Code. 15860660
Brand: VACUUBRAND™ 20696880
Only null left
Add to basket
Add to basket
Accessory for VACUUBRAND™ BVC fluid aspiration systems
  • BVC family of fluid aspiration systems deliver compact and quiet solutions for cell culture work
  • Provides flexibility to accommodate decontamination protocol whether autoclaving or disinfecting in bottle
TRUSTED_SUSTAINABILITY
Specifications
For Use With BVC Fluid Aspiration System
Type BVC Shuttle