missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BRANDplates™ pureGrade™ S 1536-well Microplates
Non-treated sterile microplates for diverse applications
Brand: BRAND™ 781702
Code : 03
Additional Details : Weight : 2.31000kg
Description
- Nontreated PS surface
- Medium binding
- Sterilized via beta-radiation
- Sterile plate for most applications
- With lid
- Available in clear, white or black
- Free from endotoxins, DNase, DNA, RNase; noncytotoxic
Bacteriological assays, screenings, storage.
Specifications
ISO 11137, AAMI | |
With Lid | |
Black | |
1536 | |
Non-treated | |
Sterile applications, bacteriological assays |
Standard | |
32 x 48 | |
Polystyrene | |
Sterile | |
Flat Bottom (F-well) | |
50/Cs. |