missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BRANDplates™ pureGrade™ S 1536-well Microplates
Non-treated sterile microplates for diverse applications
£778.21 - £820.78
Specifications
Certifications/Compliance | ISO 11137, AAMI |
---|---|
Format | Standard |
Lid | With Lid |
Array | 32 x 48 |
Material | Polystyrene |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
10688463
|
BRAND™
781700 |
Clear |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10183563
|
BRAND™
781701 |
White |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10513244
|
BRAND™
781702 |
Black |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Nontreated PS surface
- Medium binding
- Sterilized via beta-radiation
- Sterile plate for most applications
- With lid
- Available in clear, white or black
- Free from endotoxins, DNase, DNA, RNase; noncytotoxic
Bacteriological assays, screenings, storage.
Specifications
ISO 11137, AAMI | |
With Lid | |
Polystyrene | |
Sterile | |
Flat Bottom (F-well) | |
50/Cs. |
Standard | |
32 x 48 | |
1536 | |
Non-treated | |
Sterile applications, bacteriological assays |