missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates™ Smooth-Walled Insert Strips

Available with two membranes PC or PET membrane

Brand:  BRAND™ 782970

Additional Details : Weight : 0.05500kg

 View more versions of this product

Product Code. 15630657

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Sterile
  • Smooth-walled
  • cellGrade™ plus surface
  • Membrane material: PC or PET material for different optical requirements and cell visibility
  • 9 mm
  • Multiple membrane pore sizes: 0.4 μm, 1.0 μm, 3.0 μm, 8.0 μm
  • Applications: Secretion studies, co-culture, migration studies, chemotaxis assays
  • Specifications

    Specifications

    8.0 μm
    PET membrane
    12/Pk.
    Sterile
    Smooth-walled Insert Strip
    24-/6-well BRANDplates™
    cellGrade plus
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers