missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brain Heart Infusion Broth

Product Code. 12648805
Change view
Click to view available options
Description:
Brain Heart Infusion Broth
Quantity:
50/Pk.
Fill Volume:
9 mL
Fill Weight:
9 ± 0.5mL
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Quantity Fill Volume Fill Weight Unit Size
12648805 Brain Heart Infusion Broth 50/Pk. 9 mL 9 ± 0.5mL Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12648805 Supplier Thermo Scientific™ Supplier No. TV5090E
  • ONLINE EXCLUSIVE PRICE
    £95.49
    £82.53 / Pack of 50
    Save £12.96
    13% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cultivate streptococci, pneumococci, meningococci and other fastidious organisms with this highly nutritious infusion medium in prepared media tubes.

Cultivate streptococci, pneumococci, meningococci and other fastidious organisms with highly nutritious Thermo Scientific™ Brain Heart Infusion Broth in prepared media tubes.

While Brain Heart Infusion Broth was designed to detect fastidious organisms, the addition of blood and antibiotics also makes Brain Heart Infusion Broth suitable for the isolation other pathogenic fungi.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Color Daffodil yellow, transparent
Content And Storage 2 to 25°C
Description Brain Heart Infusion Broth
Format Tube
Manufacturing Origin Germany
Fill Volume 9 mL
Fill Weight 9 ± 0.5mL
Packaging Type 50 tubes in a carton box
Quantity 50/Pk.

For In Vitro Diagnostic Use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.