Learn More
Thermo Scientific™ BPC ReadyMate™ to Male Opta™ Tubing Set
Product Code. 15209397
Click to view available options
Description:
BPC ReadyMate to Male Opta Tubing Set
Length (Metric):
1 m
Quantity:
1/Pk.
Unit Size:
Each
Product Code. 15209397
Brand: Thermo Scientific™ BP0050A
Minimize prep time and maximize production for media fill trials with ready-to-use media in BioProcess Containers. Multiple connections give greater flexibility.
Minimize prep time and maximize production for media fill trials with ready-to-use media in BioProcess Containers. Create a sterile union to allow additional aseptic or steam connections using the Thermo Scientific™ BPC ReadyMate™ to Male Opta Tubing Set.
  • Achieve effective process simulations, with minimized risk of secondary contamination
  • Minimize preparation time
  • Satisfy regulatory compliance
  • Save time, reduce labor costs
Multiple plug-and-play connection options, such as BPC ReadyMate to Male Opta, are available for greater flexibility.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Opta™ is a trademark or registered trademark of the Sartorius Stedim Biotech.
ReadyMate™ is a trademark or registered trademark of the gE Healthcare BioSciences AB.
TRUSTED_SUSTAINABILITY
Specifications
Color Multi
Description BPC ReadyMate to Male Opta Tubing Set
Format Bioprocess Contaners (prepared media)
Product Type Microbiology Media
Includes Tubing and connectors
Quantity 1/Pk.
Length (Metric) 1 m