Learn More
Thermo Scientific™ BPC ReadyMate™ to Female KPC Tubing Set
Product Code. 13572040
Change view
Click to view available options
Description:
BPC ReadyMate to Female KPC Tubing Set
Length (Metric):
1 m
Quantity:
1/Pk.
Unit Size:
Each
Product Code. Description Length (Metric) Quantity unitSize
13572040 BPC ReadyMate to Female KPC Tubing Set 1 m 1/Pk. Each
1 options
Product Code. 13572040 Supplier Thermo Scientific™ Supplier No. BP0040A
Only null left
Add to basket
Add to basket
Minimize prep time and maximize production for media fill trials with ready-to-use media in BPCs. Multiple connections give greater flexibility.
Minimize prep time and maximize production for media fill trials with ready-to-use media in BioProcess Containers. Create a sterile union to allow additional aseptic or steam connections using the Thermo Scientific™ BPC ReadyMate™ to Female KPC Tubing Set.
  • Achieve effective process simulations, with minimized risk of secondary contamination
  • Minimize preparation time
  • Satisfy regulatory compliance
  • Save time, reduce labor costs
Multiple plug-and-play connection options, such as BPC ReadyMate to Female KPC, are available for greater flexibility.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
ReadyMate™ is a trademark or registered trademark of the gE Healthcare BioSciences AB.
TRUSTED_SUSTAINABILITY
Specifications
Color Multi
Description BPC ReadyMate to Female KPC Tubing Set
Format Bioprocess Contaners (prepared media)
Product Type Microbiology Media
Includes Tubing and connectors
Quantity 1/Pk.
Length (Metric) 1 m