missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bovine Serum Albumin Antibody (25G7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£363.00
Specifications
| Antigen | Bovine Serum Albumin |
|---|---|
| Clone | 25G7 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:1000 |
| Applications | Western Blot, ELISA |
Description
Bovine Serum Albumin Monoclonal specifically detects Bovine Serum Albumin in Human, Bovine, Rabbit (Negative) samples. It is validated for Western Blot, ELISA.Specifications
| Bovine Serum Albumin | |
| 1.0 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Stem Cell Markers | |
| albumin, BSA, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
| ALB | |
| IgG1 κ | |
| Protein A or G purified | |
| Reacts strongly with BSA, cross-reacts about 25% with human serum albumin but shows no cross-reaction with rabbit serum albumin. |
| 25G7 | |
| Western Blot 1:100-1:2000, ELISA 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Bovine, Rabbit (Negative) | |
| 280717 | |
| BSA purified from bovine plasma and adsorbed to aluminum hydroxide gel. | |
| Primary | |
| Store at 4C in the dark. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title