missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bovine Serum Albumin Antibody (25G7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05155
This item is not returnable.
View return policy
Description
Bovine Serum Albumin Monoclonal specifically detects Bovine Serum Albumin in Human, Bovine, Rabbit (Negative) samples. It is validated for Western Blot, ELISA.
Specifications
| Bovine Serum Albumin | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:1000 | |
| ALB | |
| BSA purified from bovine plasma and adsorbed to aluminum hydroxide gel. | |
| 100 μg | |
| Stem Cell Markers | |
| 280717 | |
| Human, Bovine, Rabbit (Negative) | |
| Purified |
| Western Blot, ELISA | |
| 25G7 | |
| Unconjugated | |
| albumin, BSA, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Reacts strongly with BSA, cross-reacts about 25% with human serum albumin but shows no cross-reaction with rabbit serum albumin. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction