Learn More
JG Finneran Associates™ Flat Bottom Inserts
Designed for use in 2mL standard opening crimp top and screw thread vials. JG Finneran™ Flat Bottom Inserts are an economical alternative to the conical limited volume inserts.
Brand: JG Finneran Associates™ 4025-531
Code : 17
Additional Details : Weight : 0.54000kg
Description
- Flat Bottom Glass and Polypropylene Inserts
- With ID ring at the top of the insert for easy open-end identification
- Available salinized or pre-cleaned
Specifications
Glass | |
250 μL | |
Flat | |
1000 Pk. |