Learn More
JG Finneran Associates™ Flat Bottom Inserts
Designed for use in 2mL standard opening crimp top and screw thread vials. JG Finneran™ Flat Bottom Inserts are an economical alternative to the conical limited volume inserts.
£213.00 - £305.00
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
12307029
|
JG Finneran Associates™
4035-630 |
350 μL |
£213.00
Pack of 1000 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10177070
|
JG Finneran Associates™
4025-531 |
250 μL |
£305.00
Pack of 1000 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
Description
- Flat Bottom Glass and Polypropylene Inserts
- With ID ring at the top of the insert for easy open-end identification
- Available salinized or pre-cleaned
Specifications
Glass | |
Flat | |
1000 Pk. |