missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Bottled Media for Sartorius Membrane Filtration Units

Product Code. 10440472 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Application):
Membrane filtration method or for direct incubation, for determination of Escherichia coli and Coliforms
Membrane filtration method or for direct incubation, for determination of LactoBacilli and Pediococci and other beer spoiling organisms
Membrane filtration method or for direct incubation, for determination of Yeasts and molds
Membrane filtration method or for direct incubation, for determination of total count
Membrane filtration method or for direct incubation, for determination of wild Yeasts
Unit Size:
Pack of 4
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) unitSize
10440472 Membrane filtration method or for direct incubation, for determination of Yeasts and molds Pack of 4
11790875 Membrane filtration method or for direct incubation, for determination of wild Yeasts Pack of 4
10544951 Membrane filtration method or for direct incubation, for determination of total count Pack of 4
10069600 Membrane filtration method or for direct incubation, for determination of Escherichia coli and Coliforms Pack of 4
10475401 Membrane filtration method or for direct incubation, for determination of LactoBacilli and Pediococci and other beer spoiling organisms Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10440472 Supplier Sartorius Supplier No. 14157A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Lysine agar media for wild yeasts determination

The traditional culture media for microorganisms is agar media. This can be used for the membrane filtration method or for direct incubation. Agar media in bottles are the cost-effective alternative for casting plates.

  • 250mL bottles, 4 bottles per box
  • Economical, provides time and cost savings
  • According to international formulas
  • Consistent quality
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Manufacturing Origin Germany
Media Type Wort
Product Type Tubed Media
Sterility Sterile
Test Type Microbial Detection
Fill Volume 250 mL
For Use With (Application) Membrane filtration method or for direct incubation, for determination of Yeasts and molds
Packaging Type Bottle
Quantity 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.