missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Bottled Media for Sartorius Membrane Filtration Units
Shop All Sartorius Products
Click to view available options
For Use With (Application):
Membrane filtration method or for direct incubation, for determination of Escherichia coli and Coliforms
Membrane filtration method or for direct incubation, for determination of LactoBacilli and Pediococci and other beer spoiling organisms
Membrane filtration method or for direct incubation, for determination of Yeasts and molds
Membrane filtration method or for direct incubation, for determination of total count
Membrane filtration method or for direct incubation, for determination of wild Yeasts
Unit Size:
Pack of 4
Description
The traditional culture media for microorganisms is agar media. This can be used for the membrane filtration method or for direct incubation. Agar media in bottles are the cost-effective alternative for casting plates.
- 250mL bottles, 4 bottles per box
- Economical, provides time and cost savings
- According to international formulas
- Consistent quality
Specifications
Specifications
| Form | Liquid |
| Manufacturing Origin | Germany |
| Media Type | Wort |
| Product Type | Tubed Media |
| Sterility | Sterile |
| Test Type | Microbial Detection |
| Fill Volume | 250 mL |
| For Use With (Application) | Membrane filtration method or for direct incubation, for determination of Yeasts and molds |
| Packaging Type | Bottle |
| Quantity | 250 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction